Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AG-2/AGR2 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $499.50
Specifications
Antigen | AG-2/AGR2 |
---|---|
Concentration | 1 mg/mL |
Dilution | Immunohistochemistry 1:100-1:300, Immunohistochemistry-Paraffin 1:100-1:300, ICC/IF (Negative) |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP105933SS
![]() |
Novus Biologicals
NBP105933SS |
0.025 mL |
Each for $206.50
|
|
|||||
NBP105933
![]() |
Novus Biologicals
NBP105933 |
0.1 mL |
Each for $499.50
|
|
|||||
Description
AG-2/AGR2 Polyclonal specifically detects AG-2/AGR2 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, ICC/IF (Negative).Specifications
AG-2/AGR2 | |
Immunohistochemistry 1:100-1:300, Immunohistochemistry-Paraffin 1:100-1:300, ICC/IF (Negative) | |
Polyclonal | |
Rabbit | |
Breast Cancer, Cancer, Stem Cell Markers | |
PBS and 30% Glycerol with 0.1% Sodium Azide | |
AG-2, AG2member 17, anterior gradient 2 homolog (Xenopus laevis), anterior gradient homolog 2 (Xenopus laevis), GOB-4, hAG-2, HPC8, secreted cement gland homolog | |
AGR2 | |
IgG | |
Affinity Purified | |
Human and mouse. Predicted to react with bovine based on 100% sequence homology. |
1 mg/mL | |
Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry | |
Unconjugated | |
RUO | |
Human, Mouse | |
O95994 | |
10551 | |
Synthetic peptide made to an internal portion of human AGR2 (within residues 50-100). [Swiss-Prot# O95994] | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title