Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AG-2/AGR2 Antibody - BSA Free, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 1 publication

$206.50 - $499.50

Specifications

Antigen AG-2/AGR2
Concentration 1 mg/mL
Dilution Western Blot 1.0 ug/ml, Immunohistochemistry 1:100-1:300, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:100-1:300
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP105936SS
SDP
View Documents
Novus Biologicals
NBP105936SS
0.025 mL
Each for $206.50
Only null left
Add to Cart
 
NBP105936
SDP
View Documents
Novus Biologicals
NBP105936
0.1 mL
Each for $499.50
Only null left
Add to Cart
 
Description

Description

AG-2/AGR2 Polyclonal specifically detects AG-2/AGR2 in Human, Mouse, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

AG-2/AGR2
Western Blot 1.0 ug/ml, Immunohistochemistry 1:100-1:300, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:100-1:300
Polyclonal
Rabbit
Breast Cancer, Cancer, Stem Cell Markers
PBS and 30% Glycerol with 0.1% Sodium Azide
AG-2, AG2member 17, anterior gradient 2 homolog (Xenopus laevis), anterior gradient homolog 2 (Xenopus laevis), GOB-4, hAG-2, HPC8, secreted cement gland homolog
AGR2
IgG
Affinity Purified
20 kDa
1 mg/mL
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
RUO
Human, Mouse, Bovine
O95994
10551
Synthetic peptide made to an N-terminal portion of mouse AGR2 (within residues 1-50). [Swiss-Prot# O88312]
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.