Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AG-2/AGR2 Antibody, DyLight 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11017780V0.1ML
Description
AG-2/AGR2 Polyclonal specifically detects AG-2/AGR2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
AG-2 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
AG-2, AG2member 17, anterior gradient 2 homolog (Xenopus laevis), anterior gradient homolog 2 (Xenopus laevis), GOB-4, hAG-2, HPC8, secreted cement gland homolog | |
Rabbit | |
Affinity Purified | |
Breast Cancer, Cancer, Stem Cell Markers | |
Human | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 405 | |
50 mM Sodium Borate | |
AGR2 | |
A synthetic peptide corresponding to an internal region of human AGR2 (within residues 50-100). [UniProt# O95994] | |
0.1 mL | |
10551 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction