Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AG-2/AGR2 Antibody, DyLight 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP105933V
Description
AG-2/AGR2 Polyclonal specifically detects AG-2/AGR2 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, ICC/IF (Negative).Specifications
AG-2/AGR2 | |
Polyclonal | |
Immunohistochemistry 1:100-1:300, Immunohistochemistry-Paraffin 1:100-1:300, ICC/IF (Negative) | |
O95994 | |
AGR2 | |
Synthetic peptide made to an internal portion of human AGR2 (within residues 50-100). [Swiss-Prot# O95994] | |
0.1 mL | |
Breast Cancer, Cancer, Stem Cell Markers | |
10551 | |
Human, Mouse | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
AG-2, AG2member 17, anterior gradient 2 homolog (Xenopus laevis), anterior gradient homolog 2 (Xenopus laevis), GOB-4, hAG-2, HPC8, secreted cement gland homolog | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human and mouse. Predicted to react with bovine based on 100% sequence homology. | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction