Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AG-2/AGR2 Antibody, mFluor Violet 500 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP105933MFV500
Description
AG-2/AGR2 Polyclonal antibody specifically detects AG-2/AGR2 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), ICC/IF (Negative)Specifications
AG-2/AGR2 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin, ICC/IF (Negative) | |
AG-2, AG2member 17, anterior gradient 2 homolog (Xenopus laevis), anterior gradient homolog 2 (Xenopus laevis), GOB-4, hAG-2, HPC8, secreted cement gland homolog | |
Synthetic peptide made to an internal portion of human AGR2 (within residues 50-100). [Swiss-Prot# O95994] | |
0.1 mL | |
Breast Cancer, Cancer, Stem Cell Markers | |
10551 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction