Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ AG 205
GREENER_CHOICESDP

Catalog No. 62425 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
62-425 5 mg
62-422-5 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 62-425 Supplier Tocris Bioscience™ Supplier No. 6242/5
Add to Cart
Add to Cart
This item is not returnable. View return policy

Progesterone receptor membrane component 1 (Pgrmc1) ligand

AG 205 is a progesterone receptor membrane component 1 (Pgrmc1) ligand. Inhibits cell cycle progression and cell viability in cancer cell lines. Inhibits progesterone induced cytosolic Ca2+ increases in GnRH neurons in vitro.

Specifications

Chemical Name or Material cis-2-[[1-(4-Chlorophenyl)-1H-tetrazol-5-yl]thio]-1-(1,2,3,4,4a,9b-hexahydro-2,8-dimethyl-5H-pyrido[4,3-b]indol-5-yl)-ethanone
CAS 442656-02-2
Assay Percent Range >98%
Quantity 5 mg
Molecular Formula C22H23ClN6OS
Formula Weight Observed MW: 454.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.