Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AG 556, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Epidermal growth factor receptor (EGFR) kinase inhibitor (IC50 = 1.1 μM). Selective over ErbB2 (IC50 > 500 μM).
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 336.39 |
| Percent Purity | >99% |
| Chemical Name or Material | AG 556 |
Safety and Handling
| Recommended Storage | Desiccate at +4°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction