Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ AGA Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556736
Description
Immunogen sequence: SSPLPLVVNT WPFKNATEAA WRALASGGSA LDAVESGCAM CEREQCDGSV Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 84%.
Aspartylglucosaminidase is involved in the catabolism of N-linked oligosaccharides of glycoproteins. It cleaves asparagine from N-acetylglucosamines as one of the final steps in the lysosomal breakdown of glycoproteins. The lysosomal storage disease aspartylglycosaminuria is caused by a deficiency in the AGA enzyme. Alternatively spliced transcript variants have been identified.
Specifications
| AGA | |
| Polyclonal | |
| Unconjugated | |
| Aga | |
| Aga; AGU; Aspartylglucosaminidase; aspartylglucosylamine deaspartylase; ASRG; AW060726; GA; Glycosylasparaginase; Glycosylasparaginase alpha chain; Glycosylasparaginase beta chain; N(4)-(Beta-N-acetylglucosaminyl)-L-asparaginase; N4-(N-acetyl-beta-glucosaminyl)-L-asparagine amidase | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 175 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P20933 | |
| Aga | |
| Recombinant protein corresponding to Human AGA. Recombinant protein control fragment (Product #RP-95817). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction