Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AGA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597441
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597441 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597441 Supplier Invitrogen™ Supplier No. PA597441
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

AGA Polyclonal Antibody for Western Blot, ELISA

Aspartylglucosaminidase is involved in the catabolism of N-linked oligosaccharides of glycoproteins. It cleaves asparagine from N-acetylglucosamines as one of the final steps in the lysosomal breakdown of glycoproteins. The lysosomal storage disease aspartylglycosaminuria is caused by a deficiency in the AGA enzyme. Alternatively spliced transcript variants have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AGA
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene Aga
Gene Accession No. P20933, Q64191
Gene Alias Aga; AGU; Aspartylglucosaminidase; aspartylglucosylamine deaspartylase; ASRG; AW060726; GA; Glycosylasparaginase; Glycosylasparaginase alpha chain; Glycosylasparaginase beta chain; N(4)-(Beta-N-acetylglucosaminyl)-L-asparaginase; N4-(N-acetyl-beta-glucosaminyl)-L-asparagine amidase
Gene Symbols Aga
Host Species Rabbit
Immunogen Recombinant Human N(4)-(beta-N-acetylglucosaminyl)-L-asparaginase protein (206-346AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11593, 175
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.