Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AGAP1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP24900525UL
Description
AGAP1 Polyclonal antibody specifically detects AGAP1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
AGAP1 | |
Polyclonal | |
Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
AGAP-1, ANK repeat and PH domains 1, ArfGAP with GTPase domain, ankyrin repeat and PH domain 1, centaurin, gamma 2, centaurin-gamma-2, CENTG2, cnt-g2, GTP-binding and GTPase-activating protein 1, KIAA1099arf-GAP with GTPase, ANK repeat and PH domain-containing protein 1, MGC71657 | |
This antibody was developed against a recombinant protein corresponding to amino acids: PDERFVLMATPNLSRRNNNRNNSSGRVPTII | |
25 μL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2), 40% Glycerol | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
116987 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction