Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AGAP1 Antibody (OTI4E8), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272357H
Description
AGAP1 Monoclonal specifically detects AGAP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| AGAP1 | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| AGAP1 | |
| Full length recombinant protein of human AGAP1 (NP_055729) produced in HEK293T cells. | |
| 0.1 mL | |
| 116987.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI4E8 | |
| Western Blot, Immunohistochemistry | |
| AGAP-1, ANK repeat and PH domains 1, ArfGAP with GTPase domain, ankyrin repeat and PH domain 1, centaurin, gamma 2, centaurin-gamma-2, CENTG2, cnt-g2, GTP-binding and GTPase-activating protein 1, KIAA1099arf-GAP with GTPase, ANK repeat and PH domain-containing protein 1, MGC71657 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction