Learn More
Invitrogen™ AGAP1 Monoclonal Antibody (OTI4E8)

Description
AGAP1 Monoclonal Antibody for Western Blot, IHC (P), Flow
Specifications
Specifications
| Antigen | AGAP1 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI4E8 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | AGAP1 |
| Gene Accession No. | Q9UPQ3 |
| Gene Alias | AGAP1; AGAP-1; Arf GAP with GTP-binding protein-like, ANK repeat and PH domains 1; ArfGAP with GTPase domain, ankyrin repeat and PH domain 1; arf-GAP with GTPase, ANK repeat and PH domain-containing protein 1; centaurin, gamma 2; centaurin-gamma-2; CENTG2; cnt-g2; GGAP1; GTP-binding and GTPase-activating protein 1; Kiaa1099; mKIAA1099; R75364 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.