Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AGAP4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559996
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559996 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559996 Supplier Invitrogen™ Supplier No. PA559996
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PSVSLEFDQQ QGSVCPSESE IYEAGA Highest antigen sequence identity to the following orthologs: Mouse - 42%, Rat - 50%.

Putative GTPase-activating protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AGAP4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene AGAP4
Gene Accession No. Q96P64
Gene Alias AGAP4; AGAP-4; AGAP8; AGAP-8; ARF GTPase-activating protein; ArfGAP with GTPase domain, ankyrin repeat and PH domain 4; ArfGAP with GTPase domain, ankyrin repeat and PH domain 8; arf-GAP with GTPase, ANK repeat and PH domain-containing protein 4; arf-GAP with GTPase, ANK repeat and PH domain-containing protein 8; centaurin, gamma-like family, member 1; centaurin, gamma-like family, member 5; Centaurin-gamma-like family member 1; Centaurin-gamma-like family member 5; CTGLF1; CTGLF5; MRIP2
Gene Symbols AGAP4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human AGAP4. Recombinant protein control fragment (Product #RP-104339).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 119016
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.