Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AGAP4 Polyclonal Antibody, Rockland™
SDP

Catalog No. 501990933
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-0933 100 μg
50-199-0934 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-0933 Supplier Rockland Immunochemicals Supplier No. 200401P07
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. An immunoblot assay detects overexpressed recombinant AGAP4.

Putative GTPase-activating protein.

Specifications

Antigen AGAP4
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.04 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene AGAP4
Gene Accession No. Q96P64
Gene Alias AGAP4; AGAP-4; AGAP8; AGAP-8; ARF GTPase-activating protein; ArfGAP with GTPase domain, ankyrin repeat and PH domain 4; ArfGAP with GTPase domain, ankyrin repeat and PH domain 8; Arf-GAP with GTPase, ANK repeat and PH domain-containing protein 4; arf-GAP with GTPase, ANK repeat and PH domain-containing protein 8; centaurin, gamma-like family, member 1; centaurin, gamma-like family, member 5; Centaurin-gamma-like family member 1; Centaurin-gamma-like family member 5; CTGLF1; CTGLF5; MRIP2
Gene Symbols AGAP4
Host Species Rabbit
Immunogen Anti-AGAP4 antibody was prepared from whole rabbit serum produced by repeated immunizations with recombinant His-tagged AGAP4 protein.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 119016
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.