Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AGAP9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA548189
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA548189 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA548189 Supplier Invitrogen™ Supplier No. PA548189
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

AGAP9 Polyclonal Antibody for Western Blot

Putative GTPase-activating protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AGAP9
Applications Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene AGAP9
Gene Accession No. Q5VTM2
Gene Alias AGAP9; AGAP-9; ArfGAP with GTPase domain, ankyrin repeat and PH domain 9; Arf-GAP with GTPase, ANK repeat and PH domain-containing protein 9; bA301J7.2; centaurin, gamma-like family, member 6; centaurin-gamma-like family member 6; CTGLF6
Gene Symbols AGAP9
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human AGAP9.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 642517
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.