Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Agar, Bacteriological Grade

Catalog No. MP215017883 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215017883 250 g
ICN15017880 100 g
MP215017890 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP215017883 Supplier MP Biomedicals™ Supplier No. 0215017883
Add to Cart
Add to Cart

Specially purified for use in preparing solid culture media for microbiological and bacteriological applications.

  • From Red alga Rhodophyceae
  • Bacteriological Grade
  • White to light yellow
  • 100 mesh powder
  • CAS: 9002-18-0
  • EC No. 232-658-1
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Content And Storage 15°C to 30°C
Form Powder
Grade Bacteriological
Product Type Nutrient Agar
For Use With (Application) Used for preparing solid culture media for microbiological and bacteriological application
Quantity 250 g
Species Red Alga Rhodophyceae
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.