Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Agar, Powder, FCC, Spectrum™ Chemical
SDP

Catalog No. 18600332 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
125 g
500 g
2.5 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail

A1672, 9002-18-0

Spectrum™ Chemical Agar, Powder, FCC is a gel made from the cell walls of several species of red algae or seaweed. It appears as an odorless, tasteless, beige powder. It functions as a binder and thickener in cooking to make jellies, pudding, and custards. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 9002-18-0
Molecular Formula C14H24O9
Molecular Weight (g/mol) 336.34
InChI Key GYYDPBCUIJTIBM-UHFFFAOYNA-N
IUPAC Name 2-({4-hydroxy-3-methyl-2,6-dioxabicyclo[3.2.1]octan-8-yl}oxy)-6-(hydroxymethyl)-4-methoxyoxane-3,5-diol
SMILES COC1C(O)C(CO)OC(OC2C3COC2C(O)C(C)O3)C1O

Specifications

CAS Max % 100%
Packaging Poly Bottle
Quantity 500 g
Grade FCC
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.