Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Agarose, Genetic Technology Grade, PROMO, Thermo Scientific Chemicals

Catalog No. J6620822PM
Click to view available options
Quantity:
100 g
  • Engineered to permit recovery of active DNA after separation, with minimal inhibition of restriction and ligation enzymes
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9012-36-6
MDL Number MFCD00081294

Specifications

Physical Form Powder
Color White
Quantity 100 g
Sensitivity Hygroscopic; Moisture Sensitive
Grade Genetic Technology
Chemical Name or Material Agarose
EINECSNumber 232-731-8
TSCA Yes

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.