Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Agarose, Low Gelling Temperature, Molecular Biology Reagent, PROMO, Thermo Scientific Chemicals

Catalog No. J6631922PM
Click to view available options
Quantity:
100 g

Applications

Agarose is useful in separation of nucleic acids electrophoretically because agarose gels have larger pore sizes than cross linked acrylamide gels at low concentrations and in chromatographic separations by either beading the agarose or cross-linking it. Agarose gels are also used in immunoelectrophoresis (IEP) and double-diffusion Ouchterlony plates to demonstrate antibody-antigen reactions.

Solubility

Soluble in water.

Notes

Gel strength (1.5% w/v):>500g/cm2; Gelling temperature: <24 to 28°C; EEO: <0.12

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9012-36-6
MDL Number MFCD00081294

Specifications

Physical Form Powder
Melting Point 65°C
Color White
Quantity 100 g
Solubility Information Soluble in water.
Chemical Name or Material Agarose
EINECSNumber 232-731-8
TSCA Yes

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.