Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Agarose, Molecular biology reagent grade, MP Biomedicals™
Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
10 g
25 g
50 g
100 g
250 g
500 g
Specifications
Specifications
Physical Form | Powder |
Color | White |
Quantity | 10 g |
Grade | Molecular Biology |
Chemical Name or Material | Agarose |
Safety and Handling
EINECSNumber | 232-731-8 |
Recommended Storage | Room temperature |
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction