Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AGBL4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101548
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101548 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101548 Supplier Invitrogen™ Supplier No. PA5101548
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total AGBL4.

Metallocarboxypeptidase that mediates deglutamylation of target proteins. Catalyzes the deglutamylation of polyglutamate side chains generated by post-translational polyglutamylation in proteins such as tubulins. Also removes polyglutamates from the carboxy-terminus of target proteins such as MYLK. Mediates deglutamylation of CGAS, regulating the antiviral activity of CGAS. Acts as a long-chain deglutamylase and specifically shortens long polyglutamate chains, while it is not able to remove the branching point glutamate, a process catalyzed by AGBL5/CCP5. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen AGBL4
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene AGBL4
Gene Accession No. Q5VU57
Gene Alias AGBL4; ATP/GTP binding protein like 4; ATP/GTP binding protein-like 4; ATP/GTP-binding protein-like 4; CCP6; Cytosolic carboxypeptidase 6
Gene Symbols AGBL4
Host Species Rabbit
Immunogen A synthesized peptide derived from human AGBL4(Accession Q5VU57), corresponding to amino acid residues Y437-K487.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84871
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.