Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AGBL5/CCP5 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 155691AP150UL

Description
AGBL5 has a function in the processing of cytosolic proteins such as alpha tubulin, which is known to be modified by the removal of a C terminal tyrosine. It is expressed in the brain. There are three named isoforms.Specifications
AGBL5/CCP5 | |
Polyclonal | |
Unconjugated | |
AGBL5 | |
AGBL5, ATP/GTP binding protein like 5, CCP5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
60509 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8NDL9 | |
AGBL5 | |
AGBL5/CCP5 Fusion Protein Ag7889 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction