Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AGBL5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114022
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114022 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114022 Supplier Invitrogen™ Supplier No. PA5114022
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RGLSSTLNVG VNKKRGLRTP PKSHNGLPVS CSENTLSRAR SFSTGTSAGG For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 86%; Pig: 100%; Rabbit: 100%; Rat: 100%.

AGBL5 has a function in the processing of cytosolic proteins such as alpha tubulin, which is known to be modified by the removal of a C terminal tyrosine. It is expressed in the brain. There are three named isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AGBL5
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene AGBL5
Gene Accession No. Q8NDL9
Gene Alias 4930455N08; 9430057O19Rik; Agbl5; ATP/GTP binding protein like 5; ATP/GTP binding protein-like 5; ATP/GTP-binding protein-like 5; carboxypeptidase 5, cytosolic; CCP5; cytosolic carboxypeptidase 5; cytosolic carboxypeptidase-like protein 5; RP75
Gene Symbols AGBL5
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C terminal region of human AGBL5 (aa 601-650).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60509
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.