Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AGE Mouse anti-Mouse, Clone: 3D11, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABN1838
Description
Anti-AGE, Methylglyoxal, clone 3D11 is a highly specific mouse monoclonal antibody that targets Methylglyoxal modified proteins and has been tested in ELISA and Immunohistochemistry (Paraffin).
Specifications
AGE | |
Monoclonal | |
Please refer to lot specific datasheet. | |
Mouse | |
Protein G Purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Immunohistochemistry (Paraffin), ELISA | |
3D11 | |
Unconjugated | |
Methylglyoxal-cojugated Ovalbumin. | |
100 μg | |
Neuroscience | |
Clone 3D11 specifically reacts with methylglyoxal modified proteins. It does not react with CML, CEL, or other AGE epitopes. | |
Stable for 1 year at 2-8°C from date of receipt. | |
IgG2a λ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction