Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AGE Mouse anti-Mouse, Clone: 6C7, MilliporeSigma™

Catalog No. MABN1837MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN1837MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN1837MI Supplier MilliporeSigma Supplier No. MABN1837
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-AGE, Carboxy-Methyl Lysine, clone 6C7 is a highly specific mouse monoclonal antibody, that targets Carboxy-Methyl Lysine (CML) modified proteins and has been tested in Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen AGE
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 6C7
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Host Species Mouse
Immunogen Carboxymethyl Lysine ­modified Keyhole Limpet Hemocyanin (KLH).
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Test Specificity Clone 6C7 specifically reacts with carboxymethyl lysine modified proteins.
Target Species Mouse
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.