Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AGE Rabbit anti-Bovine, Human, Non-human Primate, Panda, FITC, Polyclonal, FabGennix

Catalog No. 502206032
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-6032 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-6032 Supplier Fabgennix Inc Supplier No. AGEFITC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

AGE Polyclonal antibody specifically detects AGE in Bovine, Human, Non-human Primate, Panda samples. It is validated for ELISA, Immunoprecipitation, Western Blot

Specifications

Antigen AGE
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate FITC
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene Alias Advanced Glycation End Product
Host Species Rabbit
Immunogen Purified glycated ribonuclease corresponding to AGE. Glycated ribonuclease (RNase) prepared by incubating RNase with glucose at 37?C for several months.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bovine, Human, Non-human Primate, Panda
Content And Storage -20° C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.