Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AGER Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Specifications

Antigen AGER
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Form Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP15904820
SDP
View Documents
Novus Biologicals
NBP15904820UL
20 μL N/A N/A N/A
NBP159048
SDP
View Documents
Novus Biologicals
NBP159048
100 μL N/A N/A N/A
Description

Description

AGER Polyclonal specifically detects AGER in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

AGER
Polyclonal
Purified
RUO
Human
Q15109
177
Synthetic peptides corresponding to AGER(advanced glycosylation end product-specific receptor) The peptide sequence was selected from the C terminal of AGER (NP_751947). Peptide sequence RIRAGNSSPGPGDPGRPGDSRPAHWGHLVAKAATPRRGEEGPRKPGGRGG.
Primary
Store at -20C. Avoid freeze-thaw cycles.
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Cancer
PBS and 2% Sucrose with 0.09% Sodium Azide
advanced glycosylation end product-specific receptor, RAGE isoform delta, RAGE isoform sRAGE-delta, Receptor for advanced glycosylation end products, receptor for advanced glycosylation end-products
AGER
IgG
Protein A purified
43 kDa
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.