Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AGER Antibody (XT-M4) [Janelia Fluor™ 525], Novus Biologicals™
SDP

Catalog No. NB527612 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB527612 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB527612 Supplier Novus Biologicals Supplier No. NBP328638JF525
Only null left
Add to Cart
Add to Cart

Human Monoclonal Antibody

AGER Monoclonal antibody specifically detects AGER in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay

Specifications

Antigen AGER
Applications Flow Cytometry, ELISA, Functional Assay
Classification Monoclonal
Clone XT-M4
Conjugate Janelia Fluor 525
Formulation 50mM Sodium Borate
Gene Alias advanced glycosylation end product-specific receptor, RAGE isoform delta, RAGE isoform sRAGE-delta, Receptor for advanced glycosylation end products, receptor for advanced glycosylation end-products
Host Species Human
Immunogen AGER/ RAGE
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 177
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.