Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AGER Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501730885 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-0885 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-0885 Supplier Proteintech Group Inc Supplier No. 163461AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The receptor for advanced glycation endproducts (RAGE) is a member of the Immunoglobin superfamily of cell surface markers. This protein is able to interact with many molecules including: advanced glycation endproducts (AGE), amphoterin, and ligands. Research has shown that the accumulation of RAGE ligands in the biological system (endothelium mononuclear phagocytes, neurons and smooth muscle cells) can lead to destructive tissue diseases such as diabetic retinopathy, amyloidoses, tumors and inflammation disorders.

Specifications

Antigen AGER
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.23 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene AGER
Gene Accession No. Q15109, Q62151, Q63495
Gene Alias AGER, RAGE
Gene Symbols AGER
Host Species Rabbit
Immunogen AGER Fusion Protein Ag9540
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11596, 177, 81722
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.