Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AGER Rabbit anti-Mouse, Clone: 5, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP29061050UL
Description
AGER Monoclonal specifically detects AGER in Mouse samples. It is validated for ELISA.Specifications
| AGER | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| ELISA | |
| 5 | |
| ELISA 1:5000-1:10000 | |
| advanced glycosylation end product-specific receptor, RAGE isoform delta, RAGE isoform sRAGE-delta, Receptor for advanced glycosylation end products, receptor for advanced glycosylation end-products | |
| Recombinant Mouse AGER protein | |
| 50 μg | |
| Cancer | |
| 177 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction