Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AGFG1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100902
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100902 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100902 Supplier Invitrogen™ Supplier No. PA5100902
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total HRB.

HRB is related to nucleoporins, a class of proteins that mediate nucleocytoplasmic transport. HRB binds the activation domain of the human immunodeficiency virus Rev protein when Rev is assembled onto its RNA target, and is required for the nuclear export of Rev-directed RNAs. The protein encoded by this gene is related to nucleoporins, a class of proteins that mediate nucleocytoplasmic transport. This encoded protein binds the Rev activation domain when Rev is assembled onto its RNA target and can significantly enhance Rev activity when overexpressed. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AGFG1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene AGFG1
Gene Accession No. P52594, Q4KLH5, Q8K2K6
Gene Alias 0710001B06Rik; AGFG1; Agfg1-ps1; AI838505; arf-GAP domain and FG repeat-containing protein 1; arf-GAP domain and FG repeats-containing protein 1; arf-GAP domain and FG repeats-containing protein 1; arf-GAP domain and FG repeat-containing protein 1; ArfGAP with FG repeats 1; AU045498; C130049H11Rik; C85612; D730048C23Rik; HIV-1 Rev binding protein; HIV-1 Rev-binding protein; HIV-1 Rev-binding protein homolog; Hrb; hRIP, Rev interacting protein; MGC116938; MGC116940; NEUG(55-78); Neurogranin; neurogranin (protein kinase C substrate, RC3); Ng; NG/RC3; Nrgn; Nucleoporin-like protein RIP; postsynaptic spine 1; protein kinase C substrate; Pss1; R75334; RAB; Rab, Rev/Rex activation domain-binding protein; RC3; rev/Rex activation domain-binding protein; Rev-interacting protein; Rip; Unknown (protein for MGC:128795)
Gene Symbols AGFG1
Host Species Rabbit
Immunogen A synthesized peptide derived from human AGFG1(Accession P52594), corresponding to amino acid residues K6-L56.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15463, 3267, 363266
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.