Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ AGFG2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554057
Description
Immunogen sequence: FGAFTNPFTA PAAQSPLPST NPFQPNGLAP GPGFGMSSAG PGFPQAVPPT GAFASSFPAP LFPPQTPLVQ QQNGSSFGDL GSAKLGQRPL SQPAGISTNP FMTGPSSSPF ASKPPTTN Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 78%.
This gene is a member of the HIV-1 Rev binding protein (HRB) family and encodes a protein with one Arf-GAP zinc finger domain, several phe-gly (FG) motifs, and four asn-pro-phe (NPF) motifs. This protein interacts with Eps15 homology (EH) domains and plays a role in the Rev export pathway, which mediates the nucleocytoplasmic transfer of proteins and RNAs. Alternatively spliced transcript variants have been described, but their biological validity has not been determined.
Specifications
AGFG2 | |
Polyclonal | |
Unconjugated | |
AGFG2 | |
A630095P14Rik; AGFG2; Arf-GAP domain and FG repeat-containing protein 2; arf-GAP domain and FG repeats-containing protein 2; ArfGAP with FG repeats 2; HIV-1 Rev-binding protein-like protein; HRBL; nucleoporin; RABR; RAB-R; Rev/Rex activation domain binding protein-related; rev/Rex activation domain-binding protein related | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3268 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O95081 | |
AGFG2 | |
Recombinant protein corresponding to Human AGFG2. Recombinant protein control fragment (Product #RP-92675). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction