Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AGFG2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988253
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8253 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8253 Supplier Bioss Supplier No. BS8073R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene is a member of the HIV-1 Rev binding protein (HRB) family and encodes a protein with one Arf-GAP zinc finger domain, several phe-gly (FG) motifs, and four asn-pro-phe (NPF) motifs. This protein interacts with Eps15 homology (EH) domains and plays a role in the Rev export pathway, which mediates the nucleocytoplasmic transfer of proteins and RNAs. Alternatively spliced transcript variants have been described, but their biological validity has not been determined.

Specifications

Antigen AGFG2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene AGFG2
Gene Accession No. O95081
Gene Alias A630095P14Rik; AGFG2; Arf-GAP domain and FG repeat-containing protein 2; arf-GAP domain and FG repeats-containing protein 2; ArfGAP with FG repeats 2; HIV-1 Rev-binding protein-like protein; Hrbl; nucleoporin; RABR; RAB-R; Rev/Rex activation domain binding protein-related; Rev/Rex activation domain-binding protein related
Gene Symbols AGFG2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human AGFG2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3268
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.