Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AGGF1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501565656
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-5656 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-5656 Supplier Bethyl Laboratories, Inc Supplier No. A303633A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

Angiogenic factor (vasculogenesis gene on 5q), VG5Q is a critical protein for the initiation and maintece of the vascular system. It shows strong expression in blood vessels and is secreted as vessel formation is initiated. Increased angiogenesis is a molecular pathogenic mechanism of Klippel-Trenaunay syndrome (KTS). Thus, VG5Q has been shown to be involved in KTS.

Specifications

Antigen AGGF1
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene AGGF1
Gene Accession No. Q8N302
Gene Alias 2010009L17Rik, 2310029P06Rik, AW112072, Peg3, VG5Q
Gene Symbols AGGF1
Host Species Rabbit
Immunogen Between 575 and 625
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55109
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.