Learn More
AGGF1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303633AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
Angiogenic factor (vasculogenesis gene on 5q), VG5Q is a critical protein for the initiation and maintece of the vascular system. It shows strong expression in blood vessels and is secreted as vessel formation is initiated. Increased angiogenesis is a molecular pathogenic mechanism of Klippel-Trenaunay syndrome (KTS). Thus, VG5Q has been shown to be involved in KTS.Specifications
AGGF1 | |
Polyclonal | |
Unconjugated | |
AGGF1 | |
2010009L17Rik, 2310029P06Rik, AW112072, Peg3, VG5Q | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55109 | |
4° C | |
Liquid |
Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q8N302 | |
AGGF1 | |
Between 575 and 625 | |
10 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.