Learn More
AGGF1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 118891AP150UL

Description
11889-1-AP was validated by KO and KD methods and the observed MW is 84 kDa.
VG5Q (Aggf1) gene encodes an angiogenic factor that promotes proliferation of endothelial cells. Mutations in this gene are associated with a susceptibility to Klippel-Trenaunay syndrome. Pseudogenes of this gene are found on chromosomes 3, 4, 10 and 16.Specifications
AGGF1 | |
Polyclonal | |
Unconjugated | |
AGGF1 | |
AGGF1, Angiogenic factor VG5Q, GPATC7, GPATCH7, HSU84971, HUS84971, hVG5Q, VG5Q | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
310005, 55109, 66549 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q7TN31, Q8N302 | |
Aggf1 | |
AGGF1 Fusion Protein Ag2497 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.