Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aggrecan Antibody (BC-3), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB600504IR
Description
Aggrecan Monoclonal specifically detects Aggrecan in Human, Mouse, Rat, Porcine, Bovine, Canine, Equine, Feline, Guinea Pig, Rabbit, Sheep samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
Aggrecan | |
Monoclonal | |
DyLight 755 | |
AGC1SEDK, aggrecan, aggrecan core protein, Cartilage-specific proteoglycan core protein, Chondroitin sulfate proteoglycan 1, Chondroitin sulfate proteoglycan core protein 1, CSPCP, CSPG1aggrecan 1, CSPGCP, large aggregating proteoglycan, MSK16AGCAN | |
ARGSV... synthetic peptide conjugate. | |
0.1 mL | |
Extracellular Matrix, Growth and Development, Mesenchymal Stem Cell Markers, Neuronal Cell Markers, Stem Cell Markers | |
176 | |
Store at 4°C in the dark. | |
IgG1 κ |
Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
BC-3 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Feline, Guinea Pig, Rabbit, Sheep | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction