Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aggrecan Monoclonal Antibody (1R11 14A6), Invitrogen™

Catalog No. AHP0012
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AHP0012 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AHP0012 Supplier Invitrogen Supplier No. AHP0012

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

Purity is >95% by SDS-PAGE. This clone can also be referred to as clone number 1R1 14A6.

Aggrecan also known as cartilage-specific proteoglycan core protein (CSPCP) or chondroitin sulfate proteoglycan 1 is a protein that is encoded by the ACAN gene. The encoded protein is an integral part of the extracellular matrix in cartilaginous tissue and it withstands compression in cartilage. The human form of the protein is 2316 amino acids long (approximately 250 kDa) and can be expressed in multiple isoforms due to alternative splicing. Aggrecan exhibits a bottlebrush structure, in which chondroitin sulfate and keratan sulfate chains are attached to an extended protein core. The synthesis and degradation of aggrecan are being investigated for their roles in cartilage deterioration during joint injury, disease, and aging. It contains three interglobular domains (IGDs), G1, G2, and G3 that are involved in aggregation, hyaluronan binding, cell adhesion, and chondrocyte apoptosis. Aggrecan fragments from articular cartilage are released into the synovial fluid at all stages of human osteoarthritis as a result of cleavage by aggrecanase and MMP activity. Antibodies BC-3 and BC-4 can be used to detect these neo epitopes of Aggrecanase (ARGSVXXX) and MMP (XXXVDIPEN), respectively.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Aggrecan
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 1R11 14A6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.5mg/mL sodium azide; pH 7.4
Gene ACAN
Gene Accession No. P16112
Gene Alias ACAN; Agc; Agc1; AGCAN; aggrecan; aggrecan 1; aggrecan 1 (chondroitin sulfate proteoglycan 1, large aggregating proteoglycan, antigen identified by monoclonal antibody A0122); aggrecan core protein; Aggrecan core protein 2; aggrecan CS2 domain; aggrecan epidermal growth factor-like domain-1; aggrecan G2 domain; aggrecan interglobular domain; aggrecan keratan sulfate domain; aggrecan, structural proteoglycan of cartilage; articular cartilage aggrecan precursor; b2b183Clo; cartilage aggregating proteoglycan; cartilage-specific proteoglycan core protein; Chondroitin sulfate proteoglycan 1; chondroitin sulfate proteoglycan core protein 1; cmd; CSPCP; CSPG1; CSPGCP; high density cartilage proteoglycan; large aggregating cartilage proteoglycan core protein; large aggregating proteoglycan; large aggregating proteoglycan, antigen; MSK16; SEDK
Gene Symbols ACAN
Host Species Mouse
Immunogen Human natural aggrecan (proteoglycan), purified from human articular cartilage.
Purification Method Protein A
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 176
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.