Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aggrecan Neoepitope Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10074350C
Description
Aggrecan Neoepitope Polyclonal specifically detects Aggrecan Neoepitope in Human, Mouse, Rat, Porcine, Bovine, Canine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| Aggrecan Neoepitope | |
| Polyclonal | |
| Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| P16112 | |
| ACAN | |
| A synthetic peptide corresponding to a region of human Aggrecan (within residues 350-400). [UniProt# P16112] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Aggrecan Neo (JSCNIT) | |
| Store at 4C in the dark. |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| AGC1SEDK, aggrecan, aggrecan core protein, Cartilage-specific proteoglycan core protein, Chondroitin sulfate proteoglycan 1, Chondroitin sulfate proteoglycan core protein 1, CSPCP, CSPG1aggrecan 1, CSPGCP, large aggregating proteoglycan, MSK16AGCAN | |
| Rabbit | |
| 61 kDa | |
| 0.1 mL | |
| Cancer, Mesenchymal Stem Cell Markers, Stem Cell Markers | |
| 176 | |
| Human, Mouse, Rat, Pig, Bovine, Canine | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction