Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aggrecan Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501729061 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-9061 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-9061 Supplier Proteintech Group Inc Supplier No. 138801AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This polyclonal antibody raised against 1-298 aa of human aggrecan reveals a band of 70 kDa, which is consistent with the molecular weight of degradation product generated by proteolysis at the aggrecanase site.

Aggrecan also known as cartilage-specific proteoglycan core protein (CSPCP) or chondroitin sulfate proteoglycan 1 is a protein that is encoded by the ACAN gene. The encoded protein is an integral part of the extracellular matrix in cartilaginous tissue and it withstands compression in cartilage. The human form of the protein is 2316 amino acids long (approximately 250 kDa) and can be expressed in multiple isoforms due to alternative splicing. Aggrecan exhibits a bottlebrush structure, in which chondroitin sulfate and keratan sulfate chains are attached to an extended protein core. The synthesis and degradation of aggrecan are being investigated for their roles in cartilage deterioration during joint injury, disease, and aging. It contains three interglobular domains (IGDs), G1, G2, and G3 that are involved in aggregation, hyaluro binding, cell adhesion, and chondrocyte apoptosis. Aggrecan fragments from articular cartilage are released into the synovial fluid at all stages of human osteoarthritis as a result of cleavage by aggrecanase and MMP activity. Antibodies BC-3 and BC-4 can be used to detect these neo epitopes of Aggrecanase (ARGSVXXX) and MMP (XXXVDIPEN), respectively.

Specifications

Antigen Aggrecan
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.26 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene ACAN
Gene Accession No. P07897, P16112, Q61282
Gene Alias ACAN, AGC1, AGCAN, aggrecan, CSPG1, CSPGCP, MSK16, SEDK
Gene Symbols ACAN
Host Species Rabbit
Immunogen Aggrecan Fusion Protein Ag4985
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11595, 176, 58968
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.