Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aggrecan Rabbit anti-Human,Mouse, Unconjugated, Polyclonal, MilliporeSigma™

Catalog No. ABT54525UMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABT54525UMI 25 μL
ABT545100MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ABT54525UMI Supplier MilliporeSigma Supplier No. ABT54525UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Aggrecan Polyclonal antibody specifically detects Aggrecan in Human,Mouse samples. It is validated for Western Blotting, Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Aggrecan
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Gene Accession No. NP_031450
Gene Alias Aggrecan core protein;Cartilage-specific proteoglycan core protein;CSPCP
Gene Symbols Acan, Agc, Agc1
Host Species Rabbit
Immunogen GST/His-tagged recombinant fragment corresponding to 150 amino acids from the C-terminal half of murine Aggrecan core protein.
Molecular Weight of Antigen ∼270 kDa observed; 221.94 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Affinity Purified
Quantity 25 μL
Research Discipline Cell Structure
Primary or Secondary Primary
Test Specificity Anti-Aggrecan is a rabbit polyclonal antibody that detects Aggrecan core protein. It targets an epitope within 150 amino acids from the C-terminal half.
Target Species Human, Mouse
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.