Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aggrecan, Rabbit anti-Rat, Polyclonal, Millipore Sigma™

Catalog No. ABT137325UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABT137325UL 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABT137325UL Supplier MilliporeSigma Supplier No. ABT137325UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Aggrecan, Cat. No. ABT1373, is a rabbit polyclonal antibody that detects Aggrecan core protein and is tested for use in Immunohistochemistry, Immunohistochemistry (Paraffin), Radioimmunoassay, and Western Blotting.

Aggrecan core protein (UniProt: P07897; also known as Cartilage-specific proteoglycan core protein, CSPCP) is encoded by the Acan (also known as Agc, Agc1) gene (GeneID: 58968) in rat. Aggrecan is a secreted proteoglycan that is a major component of extracellular matrix of cartilagenous tissues. Its major function is to resist compression in cartilage. It binds avidly to hyaluronic acid via an N-terminal globular region and may play a regulatory role in the matrix assembly of the cartilage. Aggrecan contains two globular domains, G1 and G2 that comprise the N-terminus of the proteoglycan, while another globular region, G3, makes up the C-terminus. The G1 region is located in the N-terminus of the core protein and interacts with hyaluronic acid (HA). G1 contains Link domains and thus consists of three disulfide-bonded loop structures designated as the A, B, B' motifs. The G2 region is composed of two B-like domains, but does not have the ability to interact with HA. G2 region is separated from the G1 region by the interglobular domain, which is a prominent site for proteolysis. The keratan sulfate (KS) and the chondroitin sulfate (CS) attachment domains lie between G2 and G3. Aggrecan is present in the extracellular matrix in the form of proteoglycan aggregates, in which many aggrecan molecules interact with hyaluronan and a link protein stabilizes each interaction. It is reported that aggrecan structure changes throughout the life cycle due to changes in synthetic and degradative events. Changes due to synthesis alter the structure of the chondroitin sulfate and keratan sulfate chains, whereas those due to degradation cause cleavage of all components of the aggregate. Aggrecan is also present in most of the retina including the outer nuclear layer, the outer plexiform layer, and the inner plexiform layer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Aggrecan
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Radioimmune Assays (RIA), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Radioimmunoassay Analysis: A representative lot detected Aggrecan in Postnatal rat brain (Milev, P., et. al. (1998). Biochem Biopys Res Commun. 247(2):207-12).Immunohistochemistry Analysis: A representative lot detected Aggrecan in Embryonic day 16 rat eye, P0, P9 and adult rat retina (Popp, S., et. al. (2004). Exp Eye Res. 79(3):351-6) and Embryonic day 16, 19 rat brain and postnatal day 7 cerebellum, embryonic day 19 spinal cord (Popp, S., et. al. (2003). Dev Dyn. 227(1):143-9).Western Blotting Analysis: A representative lot detected Aggrecan in purified proteoglycans from a PBS extract of 7-day rat brain with chondroitinase ABC treatment (Popp, S., et. al. (2003). Dev Dyn. 227(1):143-9).
Formulation Rabbit polyclonal antiserum with 0.05% sodium azide.
Gene Alias Aggrecan core protein;Cartilage-specific proteoglycan core protein;CSPCP
Gene Symbols Acan;Agc;Agc1
Host Species Rabbit
Immunogen GST-tagged recombinant fragment corresponding to 213 amino acids from the C-terminal half of rat Aggrecan.
Molecular Weight of Antigen 221.12 kDa calculated
Purification Method Unpurified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Cell Structure
Primary or Secondary Primary
Test Specificity This rabbit polyclonal antibody detects aggrecan in rat eye sections.
Target Species Rat
Content And Storage Stable for 1 year at -20°C from date of receipt.
Form Serum
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.