Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BTX™ AgilePulse™ In Vivo Adapter Box
Shop All BTX Harvard Products
Click to view available options
For Use With (Equipment):
AgilePulse Generators with SN AP-4068-ID/AP-5030-IM and Before
AgilePulse Generators with SN AP-4069-ID/AP-5031-IM and After
Description
- For connection to electroporation generators
Specifications
Specifications
For Use With (Application) | In-Vivo Drug or Gene Delivery, Gene Therapy, Protein Delivery |
For Use With (Equipment) | AgilePulse Generators with SN AP-4069-ID/AP-5031-IM and After |
Product Line | BTX Electroporation |
Product Type | Cables & Adapters |
Description | AgilePulse In Vivo Adapter Box, for generators with SN AP-4069-ID / AP-5031-IM and after |
Display | Touchscreen |
Includes | Adapter Box Only |
Sterility | Non-sterile |
Connection Type | Electrode Connector Cable |
Interface | Touchscreen |
Show More |
Intended for Research use only. Not for use in diagnostic or clinical procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction