Learn More
BTX™ AgilePulse™ In Vivo Adapter Box
Catalog No. p-7250835 Shop All BTX Harvard Products
Click to view available options
For Use With (Equipment):
AgilePulse Generators with SN AP-4068-ID/AP-5030-IM and Before
AgilePulse Generators with SN AP-4069-ID/AP-5031-IM and After
AgilePulse™ In Vivo Adapter Box for use with AgilePulse™ In Vivo system.
  • For connection to electroporation generators
Specifications
For Use With (Application) In-Vivo Drug or Gene Delivery, Gene Therapy, Protein Delivery
For Use With (Equipment) AgilePulse Generators with SN AP-4068-ID/AP-5030-IM and Before
Product Line BTX Electroporation
Product Type Electroporation accessory
Description AgilePulse In Vivo Adapter Box
Display LCD
Includes Adapter Box Only
Sterility Non-sterile
Connection Type Banana Cables
Interface Digital User
Pulse Form Square Wave
Show More Show Less
Intended for Research use only. Not for use in diagnostic or clinical procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.