Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AGK Monoclonal Antibody (9A3B1)
GREENER_CHOICE

Catalog No. PIMA534959
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA534959 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA534959 Supplier Invitrogen™ Supplier No. MA534959
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Immunogen sequence: MTVFFKTLRN HWKKTTAGLC LLTWGGHWLY GKHCDNLLRR AACQEAQVFG NQLIPPNAQV KKATVFLNPA ACKGKARTLF EKNAAPILHL SGMDVTIVKT DYEGQAKKLL ELMENTDVII VAGGDGTLQE VVTGVLRRTD EATFSKIPIG FIPLGETSSL SHTLFAESGN KVQHITDATL AIVKGETVPL DVLQIKGEKE QPVFAMTGLR WGSFRDAGVK VSKYWYLGPL KIKAAHFFST LKEWPQTHQA SISYTGPTER PPNEPEETPV QRPSLYRRIL RRLASYWAQP QDALSQEVSP EVWKDVQLST IELSITTRNN QLDPTSKEDF LNICIEPDTI SKGDFITIGS RKVRNPKLHV EGTECLQASQ CTLLIPEGAG GSFSIDSEEY EAMPVEVKLL PRKLQFFCDP RKREQMLTSP TQ.

Lipid kinase that can phosphorylate both monoacylglycerol and diacylglycerol to form lysophosphatidic acid (LPA) and phosphatidic acid (PA), respectively. Does not phosphorylate sphingosine. Overexpression increases the formation and secretion of LPA, resulting in transactivation of EGFR and activation of the downstream MAPK signaling pathway, leading to increased cell growth.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AGK
Applications ELISA, Western Blot
Classification Monoclonal
Clone 9A3B1
Concentration 1.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene AGK
Gene Accession No. Q53H12
Gene Alias 2610037M15Rik; 6720408I04Rik; acylglycerol kinase; acylglycerol kinase, mitochondrial; Agk; AI465370; CATC5; CTRCT38; FLJ10842; hAGK; hsMuLK; MTDPS10; Mulk; multiple substrate lipid kinase; multi-substrate lipid kinase; RGD1562046
Gene Symbols AGK
Host Species Mouse
Immunogen Recombinant protein (or fragment.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55750
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.