Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AGK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518786
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518786 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518786 Supplier Invitrogen™ Supplier No. PA518786
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with mouse based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.

Lipid kinase that can phosphorylate both monoacylglycerol and diacylglycerol to form lysophosphatidic acid (LPA) and phosphatidic acid (PA), respectively. Does not phosphorylate sphingosine. Overexpression increases the formation and secretion of LPA, resulting in transactivation of EGFR and activation of the downstream MAPK signaling pathway, leading to increased cell growth.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AGK
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene AGK
Gene Accession No. Q53H12
Gene Alias 2610037M15Rik; 6720408I04Rik; acylglycerol kinase; acylglycerol kinase, mitochondrial; Agk; AI465370; CATC5; CTRCT38; FLJ10842; hAGK; hsMuLK; MTDPS10; Mulk; multiple substrate lipid kinase; multi-substrate lipid kinase; RGD1562046
Gene Symbols AGK
Host Species Goat
Immunogen Synthetic peptide sequence (CDPRKREQMLTSP) corresponding to the C-terminus amino acids of AGK (aa 408-421).
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55750
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.