Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AGK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597316
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA597316 100 μL
1 options

Catalog No. PIPA597316

Supplier: Invitrogen™ PA597316

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KAAHFFSTLK EWPQTHQASI SYTGPTERPP NEPEETPVQR PSLYRRILRR LASYWAQPQD ALSQEVSPEV WKDVQLSTIE LSITTRNNQL DPTSKEDFLN ICIEPDTISK GDFITIGSRK VRNPKLHVEG TECLQASQCT LLIPEGAGGS FSIDSEEYEA MPVEVKLLPR KLQFFCDPRK REQMLTSPTQ; Positive Samples: 293T, SH-SY5Y, HepG2, HeLa, 22Rv1, BT-474, K-562, Rat brain; Cellular Location: Mitochondrion membrane.

Lipid kinase that can phosphorylate both monoacylglycerol and diacylglycerol to form lysophosphatidic acid (LPA) and phosphatidic acid (PA), respectively. Does not phosphorylate sphingosine. Overexpression increases the formation and secretion of LPA, resulting in transactivation of EGFR and activation of the downstream MAPK signaling pathway, leading to increased cell growth.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AGK
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.42 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene AGK
Gene Accession No. Q53H12
Gene Alias 2610037M15Rik; 6720408I04Rik; acylglycerol kinase; acylglycerol kinase, mitochondrial; Agk; AI465370; CATC5; CTRCT38; FLJ10842; hAGK; hsMuLK; MTDPS10; Mulk; multiple substrate lipid kinase; multi-substrate lipid kinase; RGD1562046
Gene Symbols AGK
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 233-422 of human AGK (NP_0607081).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 502749, 55750
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.