Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AGL Antibody (SR1250), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | AGL |
---|---|
Clone | SR1250 |
Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence |
Classification | Monoclonal |
Description
AGL Monoclonal antibody specifically detects AGL in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, ImmunofluorescenceSpecifications
AGL | |
Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
amylo-alpha-1, 6-glucosidase, 4-alpha-glucanotransferase, GDEamylo-1, 6-glucosidase, 4-alpha-glucanotransferase, Glycogen debrancher, glycogen debranching enzyme | |
A synthesized peptide derived from human AGL (Uniprot #: P35573) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1250 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Cancer, Endocrinology, Signal Transduction | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
178 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title