Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ AGMAT Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555311
Description
Immunogen sequence: MLGTVNPSTG ALPFQSLMVA DLGDVNVNLY NLQDSCRRIQ EAYEKIVAAG CIPLTLGGDH TITYPILQAM AKKHGPVGLL HVDAHTDTTD KALGEKLYHG Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 80%.
AGMAT (Agmatinase) hydrolyzes agmatine to putrescine and urea. It is highly expressed in liver and kidney and is also found in skeletal muscle, fetal liver, brain, testis, skin and the gastrointestinal tract.
Specifications
| AGMAT | |
| Polyclonal | |
| Unconjugated | |
| AGMAT | |
| 5033405N08Rik; AGMAT; agmatinase; agmatinase, mitochondrial; Agmatine ureohydrolase; agmatine ureohydrolase (agmatinase); AUH | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79814 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BSE5 | |
| AGMAT | |
| Recombinant protein corresponding to Human AGMAT. Recombinant protein control fragment (Product #RP-94089). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction