Learn More
Agmatine sulfate, 97%
Description
Agmatine is an intermediate in the biosynthesis of the polyamines putrescine and spermine. Agmatine binds with high affinity to α2-adrenergic receptors and imidazoline binding sites and produces blockade of the NMDA-receptor associated cation channels. Agmatine is a competitive inhibitor of nitric oxide synthase activity owing to mimicry of the natural NOS substrate arginine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Agmatine is an intermediate in the biosynthesis of the polyamines putrescine and spermine. Agmatine binds with high affinity to α2-adrenergic receptors and imidazoline binding sites and produces blockade of the NMDA-receptor associated cation channels. Agmatine is a competitive inhibitor of nitric oxide synthase activity owing to mimicry of the natural NOS substrate arginine.
Solubility
Soluble in water, nearly insoluble in alcohol
Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications
Specifications
| Melting Point | 231°C to 238°C |
| Linear Formula | H2N(CH2)4NHC(=NH)NH2·H2SO4 |
| Quantity | 1 g |
| Beilstein | 3918807 |
| Merck Index | 14,188 |
| Solubility Information | Soluble in water, nearly insoluble in alcohol |
| Formula Weight | 228.27 |
| Percent Purity | 97% |
| Chemical Name or Material | Agmatine sulfate |
Safety and Handling
| EINECSNumber | 219-617-3 |
| RTECSNumber | ME8413000 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.